Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBP1/DLM-1/DAI Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB193916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB193916 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB193916 Supplier Novus Biologicals Supplier No. NBP176854AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBP1/DLM-1/DAI Polyclonal antibody specifically detects ZBP1/DLM-1/DAI in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen ZBP1/DLM-1/DAI
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias C20orf183, chromosome 20 open reading frame 183, DAI, DLM1, DLM-1, DLM1dJ718J7.3, DNA-dependent activator of interferon regulatory factors, DNA-dependent activator of IRFs, Tumor stroma and activated macrophage protein DLM-1, ZBP1 Z-DNA binding protein 1, Z-DNA binding protein 1, Z-DNA-binding protein 1
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human ZBP1. The immunogen is located within the last 50 amino acids of ZBP1. Amino Acid Squence: TPSHSKLTPKLETMTLGN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 81030
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.