Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBP1/DLM-1/DAI Antibody (Zippy-1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB172688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172688 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172688 Supplier Novus Biologicals Supplier No. NBP280056R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZBP1/DLM-1/DAI Monoclonal antibody specifically detects ZBP1/DLM-1/DAI in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen ZBP1/DLM-1/DAI
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone Zippy-1
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias C20orf183, chromosome 20 open reading frame 183, DAI, DLM1, DLM-1, DLM1dJ718J7.3, DNA-dependent activator of interferon regulatory factors, DNA-dependent activator of IRFs, Tumor stroma and activated macrophage protein DLM-1, ZBP1 Z-DNA binding protein 1, Z-DNA binding protein 1, Z-DNA-binding protein 1
Host Species Mouse
Immunogen Recombinant mouse ZBP1/DLM-1/DAI (aa 1-411)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 81030
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.