Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | ZBTB10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-4676
![]() |
Bethyl Laboratories, Inc
A303258AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
RINZF is a novel member of the Cys2-His2 zinc finger gene family, having a cDNA sequence of 3.8 kilobases. Apart from the Cys2-His2 zinc finger domains, the RIN ZF coding sequence bears little resemblance to other reported zinc finger genes. The amino-terminal region of the protein is an activation domain, despite the lack of recognizable activation motifs, such as homopolymeric glutamine and proline-rich stretches. Instead, it possesses several regions rich in basic residues, often associated with repressor domains. The gene encoding the RINZF protein is mapped on human chromosome 8q13-q21.1. The protein is highly conserved and acts as a transcriptional regulator. The protein competes with Sp1 binding at the CACC cis-regulatory element of the gastrin promoter and thus has a down-regulatory effect on gastrin transcription. The existence of two transcript variants, isoform 1 and 2, produced by alternate splicing of the RINZF gene has been studied.Specifications
ZBTB10 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
RINZF, zinc finger and BTB domain-containing protein 10, zinc finger protein RIN ZF, zinc finger protein RINZF | |
ZBTB10 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96DT7 | |
65986 | |
Between 797 and 847 | |
Primary | |
4° C | |
ZBTB10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title