Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224660 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224660 0.1 mg
NBP224660SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224660 Supplier Novus Biologicals Supplier No. NBP224660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB2 Polyclonal specifically detects ZBTB2 in Human, Mouse, Rat, Canine, Equine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZBTB2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q8N680
Gene Alias bA351K16.2, zinc finger and BTB domain containing 2, zinc finger and BTB domain-containing protein 2, ZNF437KIAA1483
Gene Symbols ZBTB2
Host Species Rabbit
Immunogen A portion of amino acids 450 to 500 of human ZBTB2 was used as immunogen for the antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57621
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoform CRA_a and CRA_b) and chimpanzee, 88% homologous in horse and cow, 82% homologous in rat and 76% homologous in mouse.
Target Species Human, Mouse, Rat, Canine, Equine, Opossum, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.