Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB24 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215165 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215165 Supplier Novus Biologicals Supplier No. NBP215165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB24 Polyclonal specifically detects ZBTB24 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZBTB24
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Alias zinc finger and BTB domain-containing protein 24, BIF1, ICF2, PATZ2, zinc finger and BTB domain containing 24, ZNF450
Gene Symbols ZBTB24
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZBTB24. The exact sequence is proprietary.
Molecular Weight of Antigen 78 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9841
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.