Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB37 Antibody, Novus Biologicals™
SDP

Catalog No. NB419141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB419141 25ul
NBP181151 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB419141 Supplier Novus Biologicals Supplier No. NBP18115125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB37 Polyclonal specifically detects ZBTB37 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ZBTB37
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias D430004I08Rik, MGC2629, zinc finger and BTB domain containing 37, zinc finger and BTB domain-containing protein 37, ZNF908
Gene Symbols ZBTB37
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PEESTSPQIIEPSSDVESREPILRINRAGQWYVETGVADRGGRSDDEVRVLGAVHIKTENLEEWLGPENQPSGEDGSSA
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84614
Test Specificity Specificity of human ZBTB37 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Mouse, Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.