Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB38 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982988
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2988 Supplier Bioss Supplier No. BS13572R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB38 is a methyl CpG binding protein that binds to DNA sequences containing a single methylated CpG, thereby acting as a transcriptional repressor. It has been found to be highly expressed in the brain and may be involved in the differentiation of late postmitotic neurons.

Specifications

Antigen ZBTB38
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZBTB38
Gene Accession No. Q8NAP3
Gene Alias A930014K01Rik; CIBZ; CtBP-interacting BTB zinc finger protein; FLJ22332; FLJ31131; FLJ35036; PPP1R171; protein phosphatase 1, regulatory subunit 171; RGD1310136; UniProtKB:Q8BW71}; ZBTB38; zbtb38 {ECO:0000250; Zenon; Zenon homolog; zinc finger and BTB domain containing 38; Zinc finger and BTB domain-containing protein 38; Zinc finger protein expressed in neurons; zinc finger transcription factor Kaiso; ZNF921
Gene Symbols ZBTB38
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZBTB38.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253461
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.