Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB40 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017179 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017179 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017179 Supplier Novus Biologicals Supplier No. NBP281868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB40 Polyclonal specifically detects ZBTB40 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZBTB40
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 5 ug/mL
Formulation PBS
Gene Alias KIAA0478ZNF923, MGC133098, zinc finger and BTB domain containing 40, zinc finger and BTB domain-containing protein 40
Gene Symbols ZBTB40
Host Species Rabbit
Immunogen ZBTB40 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human ZBTB40. The immunogen is located within amino acids 180 - 230 of ZBTB40. Amino Acid Squence: PTAQESQRNAETPAETPTT
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9923
Test Specificity ZBTB40 antibody is human specific. At least two isoforms of ZBTB40 are known to exist. This antibody is predicted to not cross-react with other ZBTB protein family members.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.