Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB46 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227211SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227211SS 0.025 mg
NBP227211 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227211SS Supplier Novus Biologicals Supplier No. NBP227211SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

ZBTB46 Polyclonal specifically detects ZBTB46 in Human samples. It is validated for ELISA.

Specifications

Antigen ZBTB46
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:1000
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias BTB (POZ) domain containing 4, BTB/POZ domain-containing protein 4, BTBD4, dJ583P15.7, dJ583P15.8, FLJ13502, RINZF, zinc finger and BTB domain containing 46, zinc finger and BTB domain-containing protein 46, Zinc finger protein 340, ZNF340
Gene Symbols ZBTB46
Host Species Rabbit
Immunogen Amino acids 62-87 of human RINZF protein were used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140685
Test Specificity The NCBI accession number forRINZF is Q96DT7. The amino acid sequence used as immunogen is 100% homologous in human and chimp and 88% homologous in mouse.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.