Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB46 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen ZBTB46
Concentration 0.5 mg/mL
Dilution ELISA 1:100-1:1000
Applications ELISA
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227211SS
SDP
View Documents
Novus Biologicals
NBP227211SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227211
SDP
View Documents
Novus Biologicals
NBP227211
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

ZBTB46 Polyclonal specifically detects ZBTB46 in Human samples. It is validated for ELISA.
Specifications

Specifications

ZBTB46
ELISA 1:100-1:1000
Polyclonal
Purified
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
140685
Amino acids 62-87 of human RINZF protein were used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
ELISA
Unconjugated
Rabbit
Human
BTB (POZ) domain containing 4, BTB/POZ domain-containing protein 4, BTBD4, dJ583P15.7, dJ583P15.8, FLJ13502, RINZF, zinc finger and BTB domain containing 46, zinc finger and BTB domain-containing protein 46, Zinc finger protein 340, ZNF340
ZBTB46
IgG
Protein G purified
The NCBI accession number forRINZF is Q96DT7. The amino acid sequence used as immunogen is 100% homologous in human and chimp and 88% homologous in mouse.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.