Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB7A/Pokemon Antibody, Novus Biologicals™
SDP

Catalog No. NBP223673 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223673 0.1 mL
NBP223673SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223673 Supplier Novus Biologicals Supplier No. NBP223673
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB7A/Pokemon Polyclonal specifically detects ZBTB7A/Pokemon in Human, Mouse, Rat, Chicken, Opossum, Chimpanzee samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Simple Western (Negative).

Specifications

Antigen ZBTB7A/Pokemon
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1: 1000, Immunohistochemistry-Paraffin 1: 100
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Alias DKFZp547O146, Factor binding IST protein 1, Factor that binds to inducer of short transcripts protein 1, FBI-1POK erythroid myeloid ontogenic factor, FBI1TTF-I-interacting peptide 21, HIV-1 inducer of short transcripts binding protein, Leukemia/lymphoma-related factor, LRFHIV-1 1st-binding protein 1, lymphoma related factor, MGC99631, pokemon, TIP21, ZBTB7zinc finger and BTB domain containing 7, zinc finger and BTB domain containing 7A, zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcriptsbinding protein, zinc finger and BTB domain-containing protein 7A, Zinc finger protein 857A, ZNF857APOZ and Krueppel erythroid myeloid ontogenic factor
Gene Symbols ZBTB7A
Host Species Rabbit
Immunogen A portion of amino acid 1-50 of human Pokemon was used as the immunogen for this antibody.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51341
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 95% homologous in mouse, rat, cow, horse and opossum, 86% homologous in dog and 77% homologous in chicken.
Target Species Human, Mouse, Rat, Chicken, Opossum, Chimpanzee
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.