Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZC3H12C Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB048880 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB048880 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB048880 Supplier Novus Biologicals Supplier No. NBP297880AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZC3H12C Polyclonal specifically detects ZC3H12C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZC3H12C
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias KIAA1726, MCP induced protein 3, MCP-induced protein 3, MCPIP3, probable ribonuclease ZC3H12C, zinc finger CCCH domain-containing protein 12C, zinc finger CCCH-type containing 12C
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ZC3H12C fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85463
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.