Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZC3H12C Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167175 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167175 100 μg
NB167176 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167175 Supplier Novus Biologicals Supplier No. NBP316040100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZC3H12C Polyclonal antibody specifically detects ZC3H12C in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen ZC3H12C
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 50% glycerol, pH7.3
Gene Alias KIAA1726, MCP induced protein 3, MCP-induced protein 3, MCPIP3, probable ribonuclease ZC3H12C, zinc finger CCCH domain-containing protein 12C, zinc finger CCCH-type containing 12C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 735-883 of human ZC3H12C (NP_203748.1). KAPHLGRSLVATRIDSISDSRLYDSSPSRQRKPYSRQEGLGSWERPGYGIDAYGYRQTYSLPDNSTQPCYEQFTFQSLPEQQEPAWRIPYCGMPQDPPRYQDNREKIYINLCNIFPPDLVRIVMKRNPHMTDAQQLAAAILVEKSQLGY
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 85463
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.