Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZC3H12D Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB189651 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB189651 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB189651 Supplier Novus Biologicals Supplier No. NBP281878B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZC3H12D Polyclonal antibody specifically detects ZC3H12D in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen ZC3H12D
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias C6orf95, FLJ46041, MCP-induced protein 4, p34, TFL, transformed follicular lymphoma, tumor suppressor TFL, zinc finger CCCH-type containing 12D
Host Species Rabbit
Immunogen ZC3H12D antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZC3H12D. The immunogen is located within amino acids 280 - 330 of ZC3H12D. Amino Acid Squence: TGARPGAGAEEQRPPRAPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 340152
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.