Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZC3H13 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292447 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292447 Supplier Novus Biologicals Supplier No. NB1002276PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZC3H13 Polyclonal specifically detects ZC3H13 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen ZC3H13
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias FLJ35669, KIAA0853DKFZp434D1812, zinc finger CCCH domain-containing protein 13, zinc finger CCCH-type containing 13
Host Species Rabbit
Immunogen Maps to a region between residue 1175 and 1225 of human Zinc Finger CCCH-type Containing 13 using the numbering given in entry NP_055885.2 (GeneID 23091).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23091
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.