Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZC3H8 Antibody (OTI4H5), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N274908A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274908A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274908A647 Supplier Novus Biologicals Supplier No. NBP274908AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZC3H8 Monoclonal specifically detects ZC3H8 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen ZC3H8
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI4H5
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Fliz1, ZC3HDC8, zinc finger CCCH domain-containing protein 8, zinc finger CCCH type domain containing 8, zinc finger CCCH-type containing 8, zinc finger CCCH-type domain containing 8
Gene Symbols ZC3H8
Host Species Mouse
Immunogen Full length recombinant protein of human ZC3H8 (NP_115883) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84524.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.