Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZCCHC11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZCCHC11 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-554-585
![]() ![]() |
Proteintech Group Inc
189801AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specifically to the 185 kDa ZCCHCC11 protein.
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002).Specifications
ZCCHC11 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
B2RX14, Q5TAX3 | |
230594, 23318 | |
Peptide | |
Primary | |
-20°C | |
ZCCHC11 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0191, PAPD3, Terminal uridylyltransferase 4, TUT4, TUTase 4, ZCCHC11 | |
Zcchc11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title