Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZCCHV Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB356347 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356347 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356347 Supplier Novus Biologicals Supplier No. NBP177184PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZCCHV Polyclonal antibody specifically detects ZCCHV in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ZCCHV
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias FLB6421DKFZp686O19171, FLJ13288antiviral 1, ZAPDKFZp686F2052, ZC3HDC2DKFZp686H1869, Zinc finger antiviral protein, Zinc finger CCCH domain-containing protein 2, zinc finger CCCH-type antiviral protein 1, zinc finger CCCH-type, antiviral 1
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human ZC3HAV1. The immunogen is located within amino acids 680 - 730 of ZC3HAV1. Amino Acid Squence: QMKRGPDHQPAKTS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 56829
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.