Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC11/ZDHHC11B Antibody, Novus Biologicals™
SDP

Catalog No. NB397259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397259 25 μL
NBP234089 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397259 Supplier Novus Biologicals Supplier No. NBP23408925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZDHHC11/ZDHHC11B Polyclonal specifically detects ZDHHC11/ZDHHC11B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZDHHC11/ZDHHC11B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P0C7U3
Gene Alias DHHC-11B, ZDHHC11B, zinc finger, DHHC-type containing 11B
Gene Symbols ZDHHC11B
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: MDTRSGSQCSVTPEAIRNNEELVLPPRISRVNGWSLP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653082
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.