Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC13 Antibody, Novus Biologicals™
SDP

Catalog No. NBP151920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151920 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151920 Supplier Novus Biologicals Supplier No. NBP151920
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ZDHHC13 Polyclonal specifically detects ZDHHC13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen ZDHHC13
Applications Western Blot, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_061901.2
Gene Alias DHHC-13, EC 2.3.1, EC 2.3.1.-, FLJ10852, FLJ10941, HIP14LHIP14-related protein, HIP3RP, huntingtin interacting protein HIP3RP, Huntingtin-interacting protein 14-related protein, Huntingtin-interacting protein HIP3RP, MGC64994, palmitoyltransferase ZDHHC13, probable palmitoyltransferase ZDHHC13, Putative MAPK-activating protein PM03, Putative NF-kappa-B-activating protein 209, Zinc finger DHHC domain-containing protein 13, zinc finger, DHHC domain containing 13, zinc finger, DHHC-type containing 13
Gene Symbols ZDHHC13
Host Species Goat
Immunogen Peptide with sequence C-FHPAREKVLRSV corresponding to C-Terminus according to NP_001001483.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54503
Test Specificity This antibody is expected to recognize both reported isoforms (NP_061901.2 and NP_001001483.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.