Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC17 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15903920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15903920 20 μL
NBP159039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15903920 Supplier Novus Biologicals Supplier No. NBP15903920UL

Rabbit Polyclonal Antibody

ZDHHC17 Polyclonal specifically detects ZDHHC17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZDHHC17
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8IUH5
Gene Alias DHHC-17, EC 2.3.1, EC 2.3.1.-, HIP14HIP-14, huntingtin interacting protein 14, huntingtin interacting protein 3, Huntingtin interacting protein H, Huntingtin yeast partner H, Huntingtin-interacting protein 14, Huntingtin-interacting protein 3, Huntingtin-interacting protein H, HYPHHIP-3, KIAA0946HIP3, palmitoyltransferase ZDHHC17, Putative MAPK-activating protein PM11, Putative NF-kappa-B-activating protein 205, Zinc finger DHHC domain-containing protein 17, zinc finger, DHHC domain containing 17, zinc finger, DHHC-type containing 17
Gene Symbols ZDHHC17
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ZDHHC17(zinc finger, DHHC-type containing 17) The peptide sequence was selected from the middle region of ZDHHC17 (NP_056151). Peptide sequence LFYNFGKSWKSDPGIIKATEEQKKKTIVELAETGSLDLSIFCSTCLIRKP.
Molecular Weight of Antigen 73 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23390
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.