Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC24 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP160079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP160079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP160079 Supplier Novus Biologicals Supplier No. NBP160079

Rabbit Polyclonal Antibody

ZDHHC24 Polyclonal specifically detects ZDHHC24 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZDHHC24
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q6UX98
Gene Alias DHHC-24, EC 2.3.1.-, probable palmitoyltransferase ZDHHC24, Zinc finger DHHC domain-containing protein 24, zinc finger, DHHC domain containing 24, zinc finger, DHHC-type containing 24
Gene Symbols ZDHHC24
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ZDHHC24(zinc finger, DHHC-type containing 24) The peptide sequence was selected from the N terminal of ZDHHC24 (NP_997223). Peptide sequence YVLVLGPGPPPLGPLARALQLALAAFQLLNLLGNVGLFLRSDPSIRGVML.
Molecular Weight of Antigen 30 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254359
Test Specificity Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Mouse: 100%; Bovine: 92%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.