Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZEB1 Antibody (639914), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB375905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375905 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375905 Supplier Novus Biologicals Supplier No. FAB6708I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZEB1 Monoclonal antibody specifically detects ZEB1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen ZEB1
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 639914
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein
Host Species Mouse
Immunogen E. coli-derived recombinant human ZEB1, Glu430-Ser575, Accession # P37275
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Immunology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6935
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.