Learn More
Description
Specifications
Specifications
| Antigen | ZEB1 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein |
| Gene Symbols | ZEB1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to an C-terminal portion of the human ZEB1 protein (between residues 1087-1124). [Uniprot: P37275] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
