Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNK4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB600284 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600284 0.1 mL
NB600284SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600284 Supplier Novus Biologicals Supplier No. NB600284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

WNK4 Polyclonal specifically detects WNK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, ICC/IF (Negative).

Specifications

Antigen WNK4
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry, Immunoprecipitation reported in scientific literature (PMID 24434184), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24434184), Immunohistochemistry-Frozen reported in scientific literature, Electron Microscopy reported in scientific literature (PMID 24434184), ICC/IF (Negative)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q96J92
Gene Alias EC 2.7.11, EC 2.7.11.1, PHA2B, PRKWNK4protein kinase, lysine deficient 4, Protein kinase lysine-deficient 4, Protein kinase with no lysine 4, serine/threonine-protein kinase WNK4, WNK lysine deficient protein kinase 4
Gene Symbols WNK4
Host Species Rabbit
Immunogen A synthetic peptide made to a region of the human WNK4 protein between residues 200-300. [UniProt# Q96J92]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Potassium Channels, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 65266
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.