Learn More
Description
Specifications
Specifications
| Antigen | WNK4 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry, Immunoprecipitation reported in scientific literature (PMID 24434184), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24434184), Immunohistochemistry-Frozen reported in scientific literature, Electron Microscopy reported in scientific literature (PMID 24434184), ICC/IF (Negative) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q96J92 |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, PHA2B, PRKWNK4protein kinase, lysine deficient 4, Protein kinase lysine-deficient 4, Protein kinase with no lysine 4, serine/threonine-protein kinase WNK4, WNK lysine deficient protein kinase 4 |
| Gene Symbols | WNK4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
