Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZEB1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP213159UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213159UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213159UV Supplier Novus Biologicals Supplier No. NBP213159UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZEB1 Polyclonal specifically detects ZEB1 in Human, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZEB1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein
Gene Symbols ZEB1
Host Species Rabbit
Immunogen A synthetic peptide made to an C-terminal portion of the human ZEB1 protein (between residues 1087-1124). [Uniprot: P37275]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Chromatin Research, Immunology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6935.0
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.