Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZEB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP16893020UL
Description
ZEB1 Polyclonal specifically detects ZEB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZEB1 | |
Polyclonal | |
Western Blot 1:100-1:2000, Immunohistochemistry | |
P37275 | |
ZEB1 | |
Synthetic peptides corresponding to ZEB1 (zinc finger E-box binding homeobox 1) The peptide sequence was selected from the N terminal of ZEB1. Peptide sequence NPRRNNVTNYNTVVETNSDSDDEDKLHIVEEESVTDAADCEGVPEDDLPT. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
PBS & 2% Sucrose. with No Preservative | |
AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein | |
Rabbit | |
124 kDa | |
20 μL | |
Cancer, Chromatin Research, Signal Transduction, Transcription Factors and Regulators, Zinc Finger | |
6935 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction