Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZEB1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $499.50

Specifications

Antigen ZEB1
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16893020
SDP
View Documents
Novus Biologicals
NBP16893020UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP168930
SDP
View Documents
Novus Biologicals
NBP168930
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

ZEB1 Polyclonal specifically detects ZEB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ZEB1
Unconjugated
RUO
P37275
6935
Synthetic peptides corresponding to ZEB1 (zinc finger E-box binding homeobox 1) The peptide sequence was selected from the N terminal of ZEB1. Peptide sequence NPRRNNVTNYNTVVETNSDSDDEDKLHIVEEESVTDAADCEGVPEDDLPT.
Primary
Polyclonal
Rabbit
Cancer, Chromatin Research, Signal Transduction, Transcription Factors and Regulators, Zinc Finger
AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein
ZEB1
IgG
124 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.