Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZEB2 Antibody (923328), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB374739 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374739 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374739 Supplier Novus Biologicals Supplier No. FAB73782I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZEB2 Monoclonal antibody specifically detects ZEB2 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen ZEB2
Applications Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 923328
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b
Host Species Mouse
Immunogen E. coli-derived recombinant human ZEB2/SIP1, Asn363-Lys537, Accession # O60315
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9839
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.