Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebrafish Basolateral Pole of Cells Antibody (FIS 2H9/1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP250370G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250370G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250370G Supplier Novus Biologicals Supplier No. NBP250370G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zebrafish Basolateral Pole of Cells Monoclonal specifically detects Zebrafish Basolateral Pole of Cells in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Zebrafish Basolateral Pole of Cells
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FIS 2H9/1
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Lysate of zebrafish intestine
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology, Epitope Tags
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.