Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebrafish Delta C Antibody (zdc2), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N250371PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N250371PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N250371PEC7 Supplier Novus Biologicals Supplier No. NBP250371PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zebrafish Delta C Monoclonal specifically detects Zebrafish Delta C in Zebrafish samples. It is validated for Flow Cytometry.

Specifications

Antigen Zebrafish Delta C
Applications Flow Cytometry
Classification Monoclonal
Clone zdc2
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Extracellular region of Zebrafish Delta C protein
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Developmental Biology, Neuroscience
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.