Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebrafish Gut Absorptive Cell Epitopes Antibody (FIS 4B7/1), HRP, Novus Biologicals™
SDP

Catalog No. NBP250373H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250373H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250373H Supplier Novus Biologicals Supplier No. NBP250373H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zebrafish Gut Absorptive Cell Epitopes Monoclonal specifically detects Zebrafish Gut Absorptive Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Zebrafish Gut Absorptive Cell Epitopes
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone FIS 4B7/1
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Host Species Mouse
Immunogen Zebrafish intestine
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.