Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebrafish Gut Absorptive Cell Epitopes Antibody (FIS 4E8/1), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309521 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309521 Supplier Novus Biologicals Supplier No. NBP250374MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zebrafish Gut Absorptive Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Absorptive Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen Zebrafish Gut Absorptive Cell Epitopes
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone FIS 4E8/1
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Lysate of zebrafish intestines
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.