Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 4B7/2), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP250376R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250376R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250376R Supplier Novus Biologicals Supplier No. NBP250376R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zebrafish Gut Secretory Cell Epitopes Monoclonal specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Zebrafish Gut Secretory Cell Epitopes
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone FIS 4B7/2
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Lysate of fish intestine
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.