Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Zenon™ Z12 Foam

Catalog No. 19148036 Shop All Protective Industrial Products Inc. Products
Click to view available options
Lens Tint:
Clear
Gray
This item is not returnable. View return policy

Catalog No. 19-148-036

Supplier: PIP™ 25001F020

This item is not returnable. View return policy

Rimless Safety Glasses feature foam padding seal and anti-scratch / anti-fog coating.

  • Lightweight, rimless design provides all day user comfort
  • Foam padding design forms a seal to keep out dust and airborne particles
  • Thin, flexible temples provide a secure fit without uncomfortable pressure points
  • Molded nose bridge design to fit the vast majority of users
  • 9.5 Base curve lens
  • Meets ANSI Z87.1+

Specifications

Lens Coating Anti-fog, Scratch-resistant
Height (Metric) 50 mm
Shape Wraparound Lens
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) General manufacturing and anywhere eye protection is needed
Frame Color Black
Temple Style Thin
Material Polycarbonate, Nylon, Synthetic Foam
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection 99.99% UVA and UVB Light Absorption
Frame Type Rimless
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.