Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zeta Opioid Receptor Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB044155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044155 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044155 Supplier Novus Biologicals Supplier No. NBP297488F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Zeta Opioid Receptor Polyclonal specifically detects Zeta Opioid Receptor in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Zeta Opioid Receptor
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias 7-60, 7-60 protein, OGFr, opioid growth factor receptor, Protein 7-60, Zeta-type opioid receptor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Zeta Opioid Receptor fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11054
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.