Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFAND2B Antibody (OTI1G10), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N274917PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274917PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274917PEC7 Supplier Novus Biologicals Supplier No. NBP274917PECY7

Mouse Monoclonal Antibody

ZFAND2B Monoclonal specifically detects ZFAND2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen ZFAND2B
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1G10
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AIRAP-like protein, AN1-type 2B, AN1-type zinc finger protein 2B, arsenite inducible RNA associated protein-like, Arsenite-inducible RNA-associated protein-like protein, zinc finger, AN1-type domain 2B
Gene Symbols ZFAND2B
Host Species Mouse
Immunogen Full length recombinant protein of human ZFAND2B (NP_620157) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 130617.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.