Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFAND3 Antibody (OTI1H10), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP274921IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274921IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274921IR Supplier Novus Biologicals Supplier No. NBP274921IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZFAND3 Monoclonal specifically detects ZFAND3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZFAND3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1H10
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AN1-type zinc finger protein 3, FLJ17799, testis expressed sequence 27, Testis-expressed sequence 27, TEX27FLJ13222, zinc finger, AN1-type domain 3
Gene Symbols ZFAND3
Host Species Mouse
Immunogen Full length recombinant protein of human ZFAND3 (NP_068762) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 60685.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.