Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFAND5 Antibody (OTI5H8), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP274922A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274922A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274922A94 Supplier Novus Biologicals Supplier No. NBP274922AF594

Mouse Monoclonal Antibody

ZFAND5 Monoclonal specifically detects ZFAND5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZFAND5
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5H8
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Zinc finger A20 domain-containing protein 2, Zinc finger protein 216ZA20D2, zinc finger, AN1-type domain 5, ZNF216A20 domain containing 2
Gene Symbols ZFAND5
Host Species Mouse
Immunogen Full length recombinant protein of human ZFAND5 (NP_005998) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7763
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.