Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFAND5 Antibody (OTI5H8), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N274922J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274922J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274922J549 Supplier Novus Biologicals Supplier No. NBP274922JF549

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

ZFAND5 Monoclonal specifically detects ZFAND5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZFAND5
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5H8
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Zinc finger A20 domain-containing protein 2, Zinc finger protein 216ZA20D2, zinc finger, AN1-type domain 5, ZNF216A20 domain containing 2
Gene Symbols ZFAND5
Host Species Mouse
Immunogen Full length recombinant protein of human ZFAND5 (NP_005998) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7763.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.