Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFHX2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$208.00 - $487.50
Specifications
| Antigen | ZFHX2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17999820
![]() |
Novus Biologicals
NBP17999820UL |
20 μL |
Each for $208.00
|
|
|||||
NBP179998
![]() |
Novus Biologicals
NBP179998 |
100 μL |
Each for $487.50
|
|
|||||
Description
ZFHX2 Polyclonal specifically detects ZFHX2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ZFHX2 | |
| Polyclonal | |
| Rabbit | |
| XP_375065 | |
| 85446 | |
| Synthetic peptide directed towards the middle region of human ZNF409. Peptide sequence GGQGSPPEASLPPSAGDKEPKTKSSWQCKVCSYETNISRNLRIHMTSEKH. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| KIAA1762, MGC141820, zinc finger homeobox 2, zinc finger homeodomain protein 2, zinc finger protein 409 | |
| ZFHX2 | |
| IgG | |
| 92 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title