Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP112 Antibody, Novus Biologicals™
SDP

Catalog No. NBP137004 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP137004 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP137004 Supplier Novus Biologicals Supplier No. NBP137004
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ZFP112 Polyclonal specifically detects ZFP112 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen ZFP112
Applications Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:200, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001076804.1
Gene Alias zfp-112, zinc finger protein 112 homolog, zinc finger protein 112 homolog (mouse), Zinc finger protein 228ZNF228
Gene Symbols ZNF112
Host Species Goat
Immunogen Peptide with sequence C-EREDDIENSHLKSYQ corresponding to internal region according to NP_001076804.1, NP_037512.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7771
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001076804.1; NP_037512.3)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.