Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP64 Antibody (PCRP-ZFP64-1H2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB385349 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385349 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385349 Supplier Novus Biologicals Supplier No. NBP308648AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZFP64 Monoclonal antibody specifically detects ZFP64 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen ZFP64
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-ZFP64-1H2
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias dJ548G19.1, dJ831D17.1, FLJ10734, FLJ12628, Zfp-64, Zinc finger protein 338MGC940, zinc finger protein 64, zinc finger protein 64 homolog (mouse), ZNF338
Host Species Mouse
Immunogen Recombinant full-length human ZFP64 protein (Uniprot: Q9NTW7)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55734
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.