Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFR Mouse anti-Human, Unconjugated, Clone: 1G2B9, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $481.52
Specifications
Antigen | ZFR |
---|---|
Clone | 1G2B9 |
Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE18752MI
|
MilliporeSigma
MABE187525UG |
25 μg |
Each for $189.29
|
|
|||||
MABE18751MI
|
MilliporeSigma
MABE1875100UG |
100 μg |
Each for $481.52
|
|
|||||
Description
ZFR Monoclonal antibody specifically detects ZFR in Human samples. It is validated for Chromatin Immunoprecipitation (ChIP), Immunohistochemistry (Paraffin), Western Blotting.Specifications
ZFR | |
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Zinc finger RNA-binding protein;hZFR;M-phase phosphoprotein homolog | |
Recombinant fragment corresponding to 372 amino acids from the C-terminal half of human Zinc finger RNA-binding protein (ZFR). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. |
1G2B9 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_057191 | |
ZFR | |
IgG1 κ | |
Protein G Purified | |
∼120 kDa observed; 117.01 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title