Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFYVE26 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176331 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176331 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176331 Supplier Novus Biologicals Supplier No. NBP176331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZFYVE26 Polyclonal specifically detects ZFYVE26 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence.

Specifications

Antigen ZFYVE26
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. EAW80952
Gene Alias DKFZp686F19106, DKFZp781H1112, FYVE domain-containing centrosomal protein, FYVE-CENT, KIAA0321zinc finger FYVE domain-containing protein 26, spastic paraplegia 15 (complicated, autosomal recessive), Spastizin, SPG15, zinc finger, FYVE domain containing 26
Gene Symbols ZFYVE26
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human SPG15. The immunogen is located within amino acids 1710 - 1760 of SPG15. Amino Acid Squence: KALDFPYPQREKRSDS
Molecular Weight of Antigen 260 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23503
Test Specificity Multiple isoforms of SPG15 are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.