Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFYVE26 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358404 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358404 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358404 Supplier Novus Biologicals Supplier No. NBP176331MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZFYVE26 Polyclonal antibody specifically detects ZFYVE26 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ZFYVE26
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp686F19106, DKFZp781H1112, FYVE domain-containing centrosomal protein, FYVE-CENT, KIAA0321zinc finger FYVE domain-containing protein 26, spastic paraplegia 15 (complicated, autosomal recessive), Spastizin, SPG15, zinc finger, FYVE domain containing 26
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human SPG15. The immunogen is located within amino acids 1710 - 1760 of SPG15. Amino Acid Squence: KALDFPYPQREKRSDS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 23503
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.