Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFYVE26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520685
Description
A suggested positive control is rat heart tissue lysate. PA5-20685 can be used with blocking peptide PEP-0801.
Hereditary spastic paraplegias (HSPs) are genetically and phenotypically heterogeneous disorders. Spastic paraplegia with thinning of the corpus callosum (ARHSP-TCC) is a relatively frequent form of complicated hereditary spastic paraplegia in which mental retardation and muscle stiffness at onset are followed by slowly progressive paraparesis and cognitive deterioration. SPG15 is the second gene known to be responsible for ARHSP-TCC in the Italian population. Mutations in this gene are associated with autosomal recessive spastic paraplegia-15. SPG15 encodes a protein containing a FYVE zinc finger binding domain which is thought to target these proteins to membrane lipids through interaction with phospholipids in the membrane. SPG15 mRNA is widely distributed in human tissues, as well as in rat embryos, suggesting a possible role for this protein during embryonic development. SPG15 co-localizes partially with endoplasmic reticulum and endosome markers, suggesting a role in intracellular trafficking. Multiple isoforms of SPG15 are known to exist.
Specifications
| ZFYVE26 | |
| Polyclonal | |
| Unconjugated | |
| ZFYVE26 | |
| 4930465A13; 9330197E15Rik; A630028O16Rik; FYVE domain-containing centrosomal protein; FYVE-CENT; Gm893; Kiaa0321; mKIAA0321; Spastizin; SPG15; Zfyve26; Zinc finger FYVE domain-containing protein 26; zinc finger FYVE-type containing 26; zinc finger, FYVE domain containing 26 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 211978, 23503, 314265 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| D4A8G9, Q5DU37, Q68DK2 | |
| ZFYVE26 | |
| A 16 amino acid peptide near the carboxy terminus of human SPG15. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction