Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFYVE27 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159421
Description
ZFYVE27 Polyclonal specifically detects ZFYVE27 in Human samples. It is validated for Western Blot.Specifications
ZFYVE27 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ32919, PROTRUDIN, RP11-459F3.2, SPG33, zinc finger FYVE domain-containing protein 27, zinc finger, FYVE domain containing 27 | |
Rabbit | |
46 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
Q5T4F4 | |
ZFYVE27 | |
Synthetic peptides corresponding to ZFYVE27(zinc finger, FYVE domain containing 27) The peptide sequence was selected from the C terminal of ZFYVE27 (NP_001002261). Peptide sequence TFSVLKKRRSCSNCGNSFCSRCCSFKVPKSSMGATAPEAQRETVFVCASC. | |
Protein A purified | |
RUO | |
118813 | |
Reconstitute in 100μL distilled water to a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction