Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFYVE28 Antibody, Novus Biologicals™
SDP

Catalog No. NB395775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395775 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395775 Supplier Novus Biologicals Supplier No. NBP24926225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZFYVE28 Polyclonal antibody specifically detects ZFYVE28 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ZFYVE28
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias hLst2, KIAA1643, lateral signaling target protein 2 homolog, LST2, LYST2, MGC43699, MGC61592, Zinc finger FYVE domain-containing protein 28, zinc finger, FYVE domain containing 28
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: ARFYYADEELNQVAAELDSLDGRKDPQRCTLLVSQFRSCQDNVLNIINQIMDECIPQDRAPRDFCVKFPEEIRHDNLAGQLWFGAECLAAGSIIMNREL
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57732
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.